Learn More
Waters Corp Waters Column, 2.5 μm, 2.1 mm x 100 mm, 130 Å
Supplier:  Waters Corp 186006103
Encompass_Preferred
These general-purpose C18 columns use Charged Surface Hybrid (CSH) technology for alternative selectivity and superior peak shape with increased loading capacity for basic compounds. Packed in ultra-low dispersion hardware, they provide exceptional separation, robustness, and throughput for HPLC assays, enabling seamless transition to UPLC.
  • Provides alternative selectivity and superior peak shape.
  • Offers exceptional separation performance and robustness.
  • Enables seamless transition to UPLC.
  • Utilizes hybrid technology and low silanol activity.
  • Designed for a pH range of 1 to 11.
  • Compatible with UPLC and UHPLC systems.
  • Enhances liquid chromatography in acidic mobile phases.
  • Ensures outstanding batch-to-batch consistency and scalability.
Catalog No. 50-179-517