Learn More
Waters Corp Waters XSelect CSH C18 XP Column, 2.5 μm, 4.6 x 30 mm, 130 Å Pore
Catalog No. 50201538 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect eXtended Performance [XP] 2.5 μm Columns are designed to enhance the transition strategy between HPLC and UPLC™ Technology. They are compatible with any LC system and provide flexibility to HPLC, UHPLC, and UPLC users to improve productivity at intermediate backpressures. These columns offer various stationary phases that are fully and seamlessly scalable between HPLC and UPLC platforms.
  • Unmatched selectivity and flexibility
    • 14 stationary phases
    • Directly scalable to HPLC and UPLC columns
    • 200+ configurations
  • Ultra-high pressure compatible, lower backpressure, improved productivity
    • Work seamlessly with any LC system
    • 40% lower backpressure
    • Superior performance
    • Higher throughput
  • Rapid method optimization
    • Methods can be transferred immediately
    • Future-proofs methods
Catalog No. 50-201-538 Supplier Waters Corp Supplier No. 186006108
XSelect eXtended Performance [XP] 2.5 μm Columns are designed to enhance the transition strategy between HPLC and UPLC™ Technology. They are compatible with any LC system and provide flexibility to HPLC, UHPLC, and UPLC users to improve productivity at intermediate backpressures. These columns offer various stationary phases that are fully and seamlessly scalable between HPLC and UPLC platforms.
  • Unmatched selectivity and flexibility
    • 14 stationary phases
    • Directly scalable to HPLC and UPLC columns
    • 200+ configurations
  • Ultra-high pressure compatible, lower backpressure, improved productivity
    • Work seamlessly with any LC system
    • 40% lower backpressure
    • Superior performance
    • Higher throughput
  • Rapid method optimization
    • Methods can be transferred immediately
    • Future-proofs methods