Learn More
Waters Corp Waters XSelect CSH C18 Column, 130Å, 2.5 μm, 3 mm × 75 mm
Catalog No. 50179259 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general purpose C18 columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
  • Offers alternative selectivity with CSH technology.
  • Achieves superior peak shape and increased loading capacity for basic compounds.
  • Provides exceptional separation performance, robustness, and throughput for HPLC.
  • Supports seamless transition to UPLC.
  • Compatible with HPLC, UHPLC, and UPLC platforms.
  • Withstands high pressures, up to 18,000 psi.
  • Enhances HPLC productivity and reduces solvent consumption.
Catalog No. 50-179-259 Supplier Waters Corp Supplier No. 176002600
XSelect CSH C18 columns are general purpose C18 columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
  • Offers alternative selectivity with CSH technology.
  • Achieves superior peak shape and increased loading capacity for basic compounds.
  • Provides exceptional separation performance, robustness, and throughput for HPLC.
  • Supports seamless transition to UPLC.
  • Compatible with HPLC, UHPLC, and UPLC platforms.
  • Withstands high pressures, up to 18,000 psi.
  • Enhances HPLC productivity and reduces solvent consumption.