Learn More
Waters Corp Waters XSelect CSH C18 Column, 2.5 μm, 2.1 mm x 100 mm, 130 Å
Catalog No. 50179256 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general purpose C18 columns featuring Charged Surface Hybrid (CSH) technology for alternative selectivity, superior peak shape, and increased loading capacity for basic compounds. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, providing exceptional separation performance, robustness, and throughput for HPLC assays and UPLC adoption.
  • Offers alternative selectivity compared to other C18 sorbents.
  • CSH technology provides superior peak shape and increased loading capacity for basic compounds.
  • Ultra-low dispersion hardware ensures exceptional separation performance.
  • Enables seamless transition to UPLC.
  • Delivers unmatched selectivity and reproducibility.
  • Overcomes column-related problems like peak shape asymmetry and poor loading.
Catalog No. 50-179-256 Supplier Waters Corp Supplier No. 176002597
XSelect CSH C18 columns are general purpose C18 columns featuring Charged Surface Hybrid (CSH) technology for alternative selectivity, superior peak shape, and increased loading capacity for basic compounds. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, providing exceptional separation performance, robustness, and throughput for HPLC assays and UPLC adoption.
  • Offers alternative selectivity compared to other C18 sorbents.
  • CSH technology provides superior peak shape and increased loading capacity for basic compounds.
  • Ultra-low dispersion hardware ensures exceptional separation performance.
  • Enables seamless transition to UPLC.
  • Delivers unmatched selectivity and reproducibility.
  • Overcomes column-related problems like peak shape asymmetry and poor loading.