Learn More
Waters Corp XSelect CSH C18 VanGuard Cartridge, 2.5 μm, 130 Å
Catalog No. 502391952 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These CSH C18 VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate contamination. They are optimized to protect analytical columns with 2.1 mm and 3.0 mm I.D. containing 2.5 μm sorbent particles, ensuring compatibility and extended analytical column life.
  • Extends analytical column lifetime and performance.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles.
  • Employs charged surface hybrid (CSH) technology for superior peak shape.
  • Enables increased mass load for preparative separation.
Catalog No. 50-239-1952 Supplier Waters Corp Supplier No. 186007817
These CSH C18 VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate contamination. They are optimized to protect analytical columns with 2.1 mm and 3.0 mm I.D. containing 2.5 μm sorbent particles, ensuring compatibility and extended analytical column life.
  • Extends analytical column lifetime and performance.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles.
  • Employs charged surface hybrid (CSH) technology for superior peak shape.
  • Enables increased mass load for preparative separation.