Learn More
Waters Corp Waters Column, PFP/Fluoro-Phenyl, 2.5 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 186006730
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl column is designed to maximize selectivity differences, providing maximum method development freedom, especially with low pH mobile phases. The non-encapped sorbent enhances retention of acidic compounds.
  • Maximizes selectivity differences
  • Offers freedom in method development, especially with low pH mobile phases
  • Non-encapped sorbent enhances retention of acidic compounds
  • Packed in ultra-low dispersion hardware
  • Delivers exceptional separation performance, robustness, and throughput for HPLC assays
  • Enables a seamless transition to UPLC adoption
  • Manufactured to stringent cGMP and ISO 9001 standards
Catalog No. 50-466-438