Learn More
Waters Corp Oasis HLB Method Validation Kit, 30 μm Particle Size
Catalog No. 50466481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB Method Validation Kit is designed to facilitate the rapid development of solid-phase extraction (SPE) methods for LC-MS analysis. It utilizes Oasis HLB, a multi-purpose, reversed-phase sorbent, which provides high capacity for a broad spectrum of compounds.
  • Designed for rapid development of SPE methods for LC-MS analysis.
  • Useful for identifying effective methods to isolate unknown analytes, zwitterionic compounds, or analyte mixtures with varied retention and elution characteristics.
  • Sorbent is water–wettable.
  • Sorbent is stable across a pH range of 0–14.
  • Sorbent is free from silanol interactions.
  • 30 μm particle size sorbent is recommended for extracting most plasma, serum, and human urine samples.
Catalog No. 50-466-481 Supplier Waters Corp Supplier No. 186006789
The Oasis HLB Method Validation Kit is designed to facilitate the rapid development of solid-phase extraction (SPE) methods for LC-MS analysis. It utilizes Oasis HLB, a multi-purpose, reversed-phase sorbent, which provides high capacity for a broad spectrum of compounds.
  • Designed for rapid development of SPE methods for LC-MS analysis.
  • Useful for identifying effective methods to isolate unknown analytes, zwitterionic compounds, or analyte mixtures with varied retention and elution characteristics.
  • Sorbent is water–wettable.
  • Sorbent is stable across a pH range of 0–14.
  • Sorbent is free from silanol interactions.
  • 30 μm particle size sorbent is recommended for extracting most plasma, serum, and human urine samples.