Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Method Validation Kit, 130Å, 2.5 μm, 4.6 mm X 150 mm

Catalog No. 50466482 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This XSelect CSH Fluoro-Phenyl method validation kit includes three columns packed with different sorbent batches, designed to maximize selectivity differences. It offers method development freedom, particularly with low pH mobile phases, and its non-encapped sorbent enhances retention of acidic compounds. These 2.5 μm eXtended Performance [XP] columns provide robust separation performance for HPLC assays and enable a smooth transition to UPLC.

  • Dimensions: 4.6 mm (Inner Diameter) × 150 mm (Length)
  • Particle size: 2.5 μm, pore size: 130 Å
  • Chemistry: PFP/Fluoro-Phenyl, separation mode: Reversed Phase
  • pH range: 1 to 8, maximum pressure: 10000 psi (690 Bar)

Catalog No. 50-466-482 Supplier Waters Corp Supplier No. 186006790
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This XSelect CSH Fluoro-Phenyl method validation kit includes three columns packed with different sorbent batches, designed to maximize selectivity differences. It offers method development freedom, particularly with low pH mobile phases, and its non-encapped sorbent enhances retention of acidic compounds. These 2.5 μm eXtended Performance [XP] columns provide robust separation performance for HPLC assays and enable a smooth transition to UPLC.

  • Dimensions: 4.6 mm (Inner Diameter) × 150 mm (Length)
  • Particle size: 2.5 μm, pore size: 130 Å
  • Chemistry: PFP/Fluoro-Phenyl, separation mode: Reversed Phase
  • pH range: 1 to 8, maximum pressure: 10000 psi (690 Bar)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.