Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Method Validation Kit, 130Å, 2.5 µm, 2.1 mm X 150 mm, 3/pk
Catalog No. 50466480 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three XSelect CSH Fluoro-Phenyl columns packed with three different batches of sorbent. Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-480 Supplier Waters Corp Supplier No. 186006788
This method validation kit consists of three XSelect CSH Fluoro-Phenyl columns packed with three different batches of sorbent. Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.