Learn More
Waters Corp Waters XSelect Column, CSH Fluoro-Phenyl, 2.5 μm, 3 mm × 150 mm
Catalog No. 50466439 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CSH Fluoro-Phenyl column optimizes selectivity differences, offering flexibility in method development, especially with low pH mobile phases. Its non-endcapped sorbent enhances retention of acidic compounds. These 2.5 μm XP columns are packed in ultra-low dispersion hardware, ensuring exceptional separation performance, robustness, and throughput for HPLC assays, and provide a smooth upgrade path to UPLC.
  • Maximizes selectivity differences; offers method development freedom, especially with low pH mobile phases.
  • Non-endcapped sorbent enhances retention of acidic compounds.
  • Packed in ultra-low dispersion hardware for exceptional separation.
  • 2.5 μm XP columns ensure robustness, high throughput for HPLC assays.
  • Enables seamless transition to UPLC.
  • Column documents are easily accessible online, reducing waste from paper COAs and eCords.
Catalog No. 50-466-439 Supplier Waters Corp Supplier No. 186006731
The CSH Fluoro-Phenyl column optimizes selectivity differences, offering flexibility in method development, especially with low pH mobile phases. Its non-endcapped sorbent enhances retention of acidic compounds. These 2.5 μm XP columns are packed in ultra-low dispersion hardware, ensuring exceptional separation performance, robustness, and throughput for HPLC assays, and provide a smooth upgrade path to UPLC.
  • Maximizes selectivity differences; offers method development freedom, especially with low pH mobile phases.
  • Non-endcapped sorbent enhances retention of acidic compounds.
  • Packed in ultra-low dispersion hardware for exceptional separation.
  • 2.5 μm XP columns ensure robustness, high throughput for HPLC assays.
  • Enables seamless transition to UPLC.
  • Column documents are easily accessible online, reducing waste from paper COAs and eCords.