Learn More
Waters Corp Column, 130Å, 5 μm, 4.6 mm x 150 mm
Catalog No. 502391477 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CSH Fluoro-Phenyl column offers high selectivity with low pH mobile phases. Its non-end-capped sorbent enhances acidic compound retention. Utilizing CSH particle technology, it improves liquid chromatography performance and delivers exceptional performance and backpressure equivalent to core-shell columns. It also provides improved loading capacity and particle size scalability.
  • Maximizes selectivity and method development freedom.
  • Enhances acidic compound retention.
  • Improves liquid chromatography performance.
  • Exceptional performance and equivalent backpressure.
  • Improved loading capacity and particle size scalability.
  • Enables seamless analytical to preparative application transition.
  • Bonded ligands redefine broadly selective phases for UHPLC separations.
Catalog No. 50-239-1477 Supplier Waters Corp Supplier No. 186005345
Add to Cart
Add to Cart
This CSH Fluoro-Phenyl column offers high selectivity with low pH mobile phases. Its non-end-capped sorbent enhances acidic compound retention. Utilizing CSH particle technology, it improves liquid chromatography performance and delivers exceptional performance and backpressure equivalent to core-shell columns. It also provides improved loading capacity and particle size scalability.
  • Maximizes selectivity and method development freedom.
  • Enhances acidic compound retention.
  • Improves liquid chromatography performance.
  • Exceptional performance and equivalent backpressure.
  • Improved loading capacity and particle size scalability.
  • Enables seamless analytical to preparative application transition.
  • Bonded ligands redefine broadly selective phases for UHPLC separations.