Learn More
Waters Corp Waters XSelect CSH Fluoro-Phenyl Column, 130 Å, 5 μm, 4.6 x 50 mm
Catalog No. 502391704 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HPLC Columns provide exceptional selectivity for chromatographic separations. XSelect CSH Fluoro-Phenyl columns use Charged Surface Hybrid (CSH™) Technology, offering alternatives to standard C18 phases, especially when selectivity for bases or planar aromatic compounds is needed. CSH Technology improves LC column performance in acidic, low ionic strength mobile phases.
  • Exceptional selectivity for chromatographic separations.
  • Offers useful alternatives to standard C18 phases.
  • Suitable for selectivity for bases or planar aromatic compounds.
  • CSH Technology improves LC column performance in acidic, low ionic strength mobile phases.
  • Improved column reproducibility and method stability.
  • Designed for method ruggedness and transferable chromatographic methods.
Catalog No. 50-239-1704 Supplier Waters Corp Supplier No. 186005342
Add to Cart
Add to Cart
The XSelect HPLC Columns provide exceptional selectivity for chromatographic separations. XSelect CSH Fluoro-Phenyl columns use Charged Surface Hybrid (CSH™) Technology, offering alternatives to standard C18 phases, especially when selectivity for bases or planar aromatic compounds is needed. CSH Technology improves LC column performance in acidic, low ionic strength mobile phases.
  • Exceptional selectivity for chromatographic separations.
  • Offers useful alternatives to standard C18 phases.
  • Suitable for selectivity for bases or planar aromatic compounds.
  • CSH Technology improves LC column performance in acidic, low ionic strength mobile phases.
  • Improved column reproducibility and method stability.
  • Designed for method ruggedness and transferable chromatographic methods.