Learn More
Waters Corp Waters Column, 130Å, 3.5 μm, 1 mm, 50 mm
Catalog No. 50791332 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed to maximize selectivity differences, providing maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides enhanced retention of acidic compounds.
  • Maximizes selectivity differences
  • Provides maximum method development freedom, especially when using low pH mobile phases
  • Non-encapped sorbent provides enhanced retention of acidic compounds
Catalog No. 50-791-332 Supplier Waters Corp Supplier No. 186005304
Add to Cart
Add to Cart
This column is designed to maximize selectivity differences, providing maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides enhanced retention of acidic compounds.
  • Maximizes selectivity differences
  • Provides maximum method development freedom, especially when using low pH mobile phases
  • Non-encapped sorbent provides enhanced retention of acidic compounds