Learn More
Waters Corp XSelect Analytical Column, Fluoro-Phenyl, 5 μm, 4.6 x 75 mm
Catalog No. 50791368 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect Analytical Column is designed with Fluoro-Phenyl chemistry, offering a 5 μm particle size. Its dimensions are 4.6 x 75 mm, making it suitable for various analytical applications.
  • Features Fluoro-Phenyl chemistry
  • Has a particle size of 5 μm
  • Dimensions are 4.6 x 75 mm
  • Utilizes Charged Surface Hybrid (CSH) particle technology
Catalog No. 50-791-368 Supplier Waters Corp Supplier No. 186005343
This XSelect Analytical Column is designed with Fluoro-Phenyl chemistry, offering a 5 μm particle size. Its dimensions are 4.6 x 75 mm, making it suitable for various analytical applications.
  • Features Fluoro-Phenyl chemistry
  • Has a particle size of 5 μm
  • Dimensions are 4.6 x 75 mm
  • Utilizes Charged Surface Hybrid (CSH) particle technology