Learn More
Waters Corp WATERS CORPORATION Column, CSH Fluoro-Phenyl, 130Å, 3.5 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186005311
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl column offers method development flexibility with low pH mobile phases and enhanced retention of acidic compounds due to its non-endcapped sorbent. It provides superior separation performance and robustness across HPLC, UPLC, and UHPLC systems, utilizing Charged Surface Hybrid (CSH) technology for optimal liquid chromatography.
  • Maximizes selectivity differences, especially with low pH mobile phases.
  • Non-endcapped sorbent for enhanced retention of acidic compounds.
  • Compatible with HPLC, UPLC, and UHPLC technologies.
  • Superior separation performance, robustness, and throughput.
  • Exceptional performance and equal back pressure to core-shell columns.
  • Utilizes CSH technology for improved liquid chromatography in acidic, low ionic strength mobile phases.
Catalog No. 50-791-339