Learn More
Waters Corp Waters XSelect CSH Fluoro-Phenyl Column, 130Å, 3.5 μm, 4.6 x 50 mm
Catalog No. 50791349 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect CSH Fluoro-Phenyl Column is designed for HPLC applications, offering exceptional selectivity and reproducibility. It features a 130Å pore diameter, 3.5 μm particle size, and 4.6 x 50 mm dimensions. CSH technology enhances performance in acidic, low ionic strength mobile phases.
  • Exceptional selectivity and reproducibility.
  • Designed for a wide range of chromatographic separations.
  • CSH technology improves LC column performance in acidic, low ionic strength mobile phases.
  • Fluorinated phases offer alternatives to standard C18 phases, ideal for selectivity of bases or planar aromatic compounds.
  • Provides superior peak shape for basic compounds and low column bleed.
  • Excellent batch-to-batch reproducibility and ultra-efficiencies.
  • Seamless transferability between different particle sizes and column formats.
Catalog No. 50-791-349 Supplier Waters Corp Supplier No. 186005322
This XSelect CSH Fluoro-Phenyl Column is designed for HPLC applications, offering exceptional selectivity and reproducibility. It features a 130Å pore diameter, 3.5 μm particle size, and 4.6 x 50 mm dimensions. CSH technology enhances performance in acidic, low ionic strength mobile phases.
  • Exceptional selectivity and reproducibility.
  • Designed for a wide range of chromatographic separations.
  • CSH technology improves LC column performance in acidic, low ionic strength mobile phases.
  • Fluorinated phases offer alternatives to standard C18 phases, ideal for selectivity of bases or planar aromatic compounds.
  • Provides superior peak shape for basic compounds and low column bleed.
  • Excellent batch-to-batch reproducibility and ultra-efficiencies.
  • Seamless transferability between different particle sizes and column formats.