Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Column, 130Å, 3.5 µm, 3 mm X 20 mm, 1/pk
Catalog No. 50791342 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-791-342 Supplier Waters Corp Supplier No. 186005314
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.