Learn More
Waters Corp Waters XSelect Fluoro-Phenyl Method Validation Kit, 5 μm, 130 Å
Catalog No. 50791513 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HPLC columns, powered by CSH Technology, offer a wide range of selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases.
  • Provide a wide range of selectivities
  • Deliver superior performance in acidic, low ionic strength, LC/MS friendly mobile phases
  • Contain three columns
  • Each packed with a different gel lot of stationary phase material
Catalog No. 50-791-513 Supplier Waters Corp Supplier No. 186005556
XSelect HPLC columns, powered by CSH Technology, offer a wide range of selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases.
  • Provide a wide range of selectivities
  • Deliver superior performance in acidic, low ionic strength, LC/MS friendly mobile phases
  • Contain three columns
  • Each packed with a different gel lot of stationary phase material