Learn More
Waters Corp Prep Column, CSH Fluoro-Phenyl, 5 μm, 10 × 100 mm
Catalog No. 502390596 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect CSH Fluoro-Phenyl OBD Prep Column is a High Performance Liquid Chromatography (HPLC) column for scaling screening methods to preparative HPLC. It uses CSH (Charged Surface Hybrid) Technology for added selectivity and low pH loadability.
  • Achieve fast, efficient, lab-scale separations for greater throughput.
  • Directly scale from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Robust chromatographic particles designed for purification.
  • Patented OBD Preparative Column design results in predictable, uniform density, eliminating voids.
  • Exceptional resistance to mechanical bed failure, extending column lifetimes.
  • Maximizes productivity by scaling screening methods to preparative HPLC with UPLC/UHPLC speed.
  • Ensures scalability from analytical to preparative dimensions by matching column bed densities.
Catalog No. 50-239-0596 Supplier Waters Corp Supplier No. 186008241
This XSelect CSH Fluoro-Phenyl OBD Prep Column is a High Performance Liquid Chromatography (HPLC) column for scaling screening methods to preparative HPLC. It uses CSH (Charged Surface Hybrid) Technology for added selectivity and low pH loadability.
  • Achieve fast, efficient, lab-scale separations for greater throughput.
  • Directly scale from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Robust chromatographic particles designed for purification.
  • Patented OBD Preparative Column design results in predictable, uniform density, eliminating voids.
  • Exceptional resistance to mechanical bed failure, extending column lifetimes.
  • Maximizes productivity by scaling screening methods to preparative HPLC with UPLC/UHPLC speed.
  • Ensures scalability from analytical to preparative dimensions by matching column bed densities.