Learn More
Waters Corp WATERS CORPORATION XSelect CSH Fluoro-Phenyl Column, 2.5 μm, 130 Å, 3 mm x 100 mm
Supplier:  Waters Corp 186006119
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl column is designed to maximize selectivity differences and provides method development freedom, especially with low pH mobile phases. Its non-encapped sorbent enhances the retention of acidic compounds. These 2.5 μm eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, offer exceptional separation performance, robustness, and throughput for HPLC assays, while also providing a seamless transition path to UPLC adoption.
  • Maximizes selectivity differences.
  • Provides method development freedom with low pH mobile phases.
  • Non-encapped sorbent for enhanced retention of acidic compounds.
  • Packed in ultra-low dispersion hardware.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Offers a seamless transition path toward UPLC adoption.
Catalog No. 50-179-528