Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Method Validation Kit, 130Å, 2.5 µm, 2.1 mm X 100 mm, 3/pk
Catalog No. 50179605 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three batches of XSelect CSH Fluoro-Phenyl columns packed with three different batches of sorbent
Catalog No. 50-179-605 Supplier Waters Corp Supplier No. 186006240
This method validation kit consists of three batches of XSelect CSH Fluoro-Phenyl columns packed with three different batches of sorbent