Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Column, 130Å, 2.5 μm, 2.1 mm × 100 mm
Catalog No. 50179264 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is part of the XSelect CSH Fluoro-Phenyl family, designed to provide enhanced separation and robust performance for various chromatographic applications. It utilizes CSH particle technology to offer superior peak shape and efficiency, making it suitable for demanding UPLC and UHPLC methods.
  • Provides reversed-phase separation with PFP/Fluoro-Phenyl chemistry.
  • Features a hybrid, spherical particle substrate with low silanol activity.
  • Offers a particle size of 2.5 μm and a pore size of 130 Å.
  • Designed for high-pressure applications, up to 18000 psi (1240 bar).
  • Compatible with a broad pH range of 1 to 8.
  • Ideal for UPLC and UHPLC systems, enhancing method development.
Catalog No. 50-179-264 Supplier Waters Corp Supplier No. 176002605
This column is part of the XSelect CSH Fluoro-Phenyl family, designed to provide enhanced separation and robust performance for various chromatographic applications. It utilizes CSH particle technology to offer superior peak shape and efficiency, making it suitable for demanding UPLC and UHPLC methods.
  • Provides reversed-phase separation with PFP/Fluoro-Phenyl chemistry.
  • Features a hybrid, spherical particle substrate with low silanol activity.
  • Offers a particle size of 2.5 μm and a pore size of 130 Å.
  • Designed for high-pressure applications, up to 18000 psi (1240 bar).
  • Compatible with a broad pH range of 1 to 8.
  • Ideal for UPLC and UHPLC systems, enhancing method development.