Learn More
Waters Corp Waters XSelect CSH Fluoro-Phenyl XP Column, 2.5 μm, 3 × 100 mm
Catalog No. 50179268 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Fluoro-Phenyl columns maximize selectivity differences and offer method development freedom, particularly with low pH mobile phases. The non-encapped sorbent enhances retention of acidic compounds. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns provide exceptional separation performance, robustness, and throughput for HPLC assays, aiding UPLC adoption.
  • Designed to maximize selectivity differences, offering maximum method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent provides enhanced retention of acidic compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm extended performance (XP) columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-179-268 Supplier Waters Corp Supplier No. 176002609
XSelect CSH Fluoro-Phenyl columns maximize selectivity differences and offer method development freedom, particularly with low pH mobile phases. The non-encapped sorbent enhances retention of acidic compounds. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns provide exceptional separation performance, robustness, and throughput for HPLC assays, aiding UPLC adoption.
  • Designed to maximize selectivity differences, offering maximum method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent provides enhanced retention of acidic compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm extended performance (XP) columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.