Learn More
Waters Corp Column, PFP/Fluoro-Phenyl, 2.5 μm, 4.6 mm, 50 mm
Catalog No. 50201543 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed to maximize selectivity differences, offering maximum method development freedom, particularly when using low pH mobile phases. Its non-encapped sorbent provides enhanced retention of acidic compounds. The 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays, while also providing a seamless transition path toward UPLC adoption.
  • Designed to maximize selectivity differences, providing maximum method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-201-543 Supplier Waters Corp Supplier No. 186006121
This column is designed to maximize selectivity differences, offering maximum method development freedom, particularly when using low pH mobile phases. Its non-encapped sorbent provides enhanced retention of acidic compounds. The 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays, while also providing a seamless transition path toward UPLC adoption.
  • Designed to maximize selectivity differences, providing maximum method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.