Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HPLC Column, Fluoro-Phenyl, 2.5 μm, 2.1 x 30 mm

Catalog No. 50179261 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect CSH fluorophenyl column maximizes selectivity differences, offering freedom in method development, especially with low pH mobile phases. Its uncapped sorbent enhances retention of acidic compounds. These 2.5 μm XP columns, with ultra-low dispersion hardware, provide exceptional separation, robustness, and high throughput for HPLC assays, facilitating seamless UPLC transition.

  • Maximizes selectivity differences and offers extensive freedom in method development.
  • Uncapped sorbent improves retention of acidic compounds.
  • Designed for exceptional separation performance, robustness, and high throughput in HPLC assays.
  • Enables a seamless transition to UPLC.
  • Inner diameter: 2.1 mm
  • Length: 30 mm
  • Particle size: 2.5 μm
  • Pore size: 130 Å
  • pH Range: 1 to 8
  • Carbon load: 10.0%
  • Mode: Reversed Phase (RP)

Catalog No. 50-179-261 Supplier Waters Corp Supplier No. 176002602
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect CSH fluorophenyl column maximizes selectivity differences, offering freedom in method development, especially with low pH mobile phases. Its uncapped sorbent enhances retention of acidic compounds. These 2.5 μm XP columns, with ultra-low dispersion hardware, provide exceptional separation, robustness, and high throughput for HPLC assays, facilitating seamless UPLC transition.

  • Maximizes selectivity differences and offers extensive freedom in method development.
  • Uncapped sorbent improves retention of acidic compounds.
  • Designed for exceptional separation performance, robustness, and high throughput in HPLC assays.
  • Enables a seamless transition to UPLC.
  • Inner diameter: 2.1 mm
  • Length: 30 mm
  • Particle size: 2.5 μm
  • Pore size: 130 Å
  • pH Range: 1 to 8
  • Carbon load: 10.0%
  • Mode: Reversed Phase (RP)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.