Learn More
Waters Corp Waters Corporation Column, Phenyl, 2.5 μm, 2.1 mm, 150 mm
Catalog No. 50466441 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns provide alternative analyte selectivity to traditional reversed-phase columns, aiding method development. They feature a robust, low bleed C6 phenyl-ligand, selectively retaining polyaromatic compounds via pi-pi interactions. Packed in ultra-low dispersion hardware, 2.5 μm XP particles offer exceptional separation, robustness, and throughput for HPLC assays, facilitating seamless UPLC transition. All column documents, including COAs, are available online.
  • Alternative analyte selectivity aids method development.
  • Robust, low bleed C6 phenyl-ligand for selective polyaromatic compound retention.
  • Exceptional separation performance from 2.5 μm XP particles.
  • Enhanced robustness and high throughput for HPLC assays.
  • Seamless transition path towards UPLC adoption.
  • Online access to all column documents, including COAs.
Catalog No. 50-466-441 Supplier Waters Corp Supplier No. 186006733
These columns provide alternative analyte selectivity to traditional reversed-phase columns, aiding method development. They feature a robust, low bleed C6 phenyl-ligand, selectively retaining polyaromatic compounds via pi-pi interactions. Packed in ultra-low dispersion hardware, 2.5 μm XP particles offer exceptional separation, robustness, and throughput for HPLC assays, facilitating seamless UPLC transition. All column documents, including COAs, are available online.
  • Alternative analyte selectivity aids method development.
  • Robust, low bleed C6 phenyl-ligand for selective polyaromatic compound retention.
  • Exceptional separation performance from 2.5 μm XP particles.
  • Enhanced robustness and high throughput for HPLC assays.
  • Seamless transition path towards UPLC adoption.
  • Online access to all column documents, including COAs.