Learn More
Waters Corp Waters XSelect CSH Phenyl-Hexyl Kit, 2.5 μm, 4.6 mm × 150 mm
Catalog No. 50466485 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit, featuring three columns with different sorbent batches, offers alternative analyte selectivity for method development. Its trifunctionally bonded C6 phenyl-ligand provides a robust, low-bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions. The 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance and throughput for HPLC assays, enabling a seamless transition to UPLC.
  • Three columns packed with different sorbent batches.
  • Alternative analyte selectivity for method development.
  • Trifunctionally bonded C6 phenyl-ligand ensures robust, low-bleed performance.
  • Selectively retains polyaromatic compounds via pi-pi interactions.
  • 2.5 μm XP columns provide exceptional separation performance and throughput for HPLC.
  • Seamless transition path to UPLC adoption.
Catalog No. 50-466-485 Supplier Waters Corp Supplier No. 186006793
This method validation kit, featuring three columns with different sorbent batches, offers alternative analyte selectivity for method development. Its trifunctionally bonded C6 phenyl-ligand provides a robust, low-bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions. The 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance and throughput for HPLC assays, enabling a seamless transition to UPLC.
  • Three columns packed with different sorbent batches.
  • Alternative analyte selectivity for method development.
  • Trifunctionally bonded C6 phenyl-ligand ensures robust, low-bleed performance.
  • Selectively retains polyaromatic compounds via pi-pi interactions.
  • 2.5 μm XP columns provide exceptional separation performance and throughput for HPLC.
  • Seamless transition path to UPLC adoption.