Learn More
Waters Corp XSelect Method Validation Kit, 2.5 μm, 2.1 mm, 150 mm, 3/pk
Catalog No. 50466483 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns, each packed with a different batch of sorbent. These columns provide an alternative analyte selectivity compared to traditional reversed-phase columns, proving valuable for method development. They feature a trifunctionally bonded C6 phenyl-ligand, ensuring a robust, low bleed sorbent that selectively retains polyaromatic compounds.
  • Offers alternative analyte selectivity for method development.
  • Utilizes a trifunctionally bonded C6 phenyl-ligand.
  • Provides robust, low bleed sorbent with π–π interactions.
  • Features 2.5 μm XP columns in ultra-low dispersion hardware.
  • Ensures exceptional separation performance and robustness.
  • Supports high throughput for HPLC assays and UPLC adoption.
  • Digital access to all documentation, including Certificates of Analysis.
Catalog No. 50-466-483 Supplier Waters Corp Supplier No. 186006791
This method validation kit includes three columns, each packed with a different batch of sorbent. These columns provide an alternative analyte selectivity compared to traditional reversed-phase columns, proving valuable for method development. They feature a trifunctionally bonded C6 phenyl-ligand, ensuring a robust, low bleed sorbent that selectively retains polyaromatic compounds.
  • Offers alternative analyte selectivity for method development.
  • Utilizes a trifunctionally bonded C6 phenyl-ligand.
  • Provides robust, low bleed sorbent with π–π interactions.
  • Features 2.5 μm XP columns in ultra-low dispersion hardware.
  • Ensures exceptional separation performance and robustness.
  • Supports high throughput for HPLC assays and UPLC adoption.
  • Digital access to all documentation, including Certificates of Analysis.