Learn More
Waters Corp Waters XSelect CSH Phenyl-Hexyl Column, 130 Å, 2.5 μm
Catalog No. 50466442 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development, utilizing a robust, low bleed C6 phenyl-ligand for selective retention of polyaromatic compounds via π–π interactions. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns offer exceptional separation performance, robustness, and throughput for HPLC assays, enabling a seamless transition to UPLC. Optimized for low pH mobile phases, the non-end-capped sorbent enhances acidic chemical retention.
  • Emphasizes reversed-phase contacts over secondary Lewis-base interactions.
  • Incorporates Charged Surface Hybrid (CSH) technology for enhanced liquid chromatography performance in acidic, low ionic strength mobile phases.
  • Offers increased loading capacity and particle size scalability, typical of fully-porous particle columns.
Catalog No. 50-466-442 Supplier Waters Corp Supplier No. 186006734
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development, utilizing a robust, low bleed C6 phenyl-ligand for selective retention of polyaromatic compounds via π–π interactions. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns offer exceptional separation performance, robustness, and throughput for HPLC assays, enabling a seamless transition to UPLC. Optimized for low pH mobile phases, the non-end-capped sorbent enhances acidic chemical retention.
  • Emphasizes reversed-phase contacts over secondary Lewis-base interactions.
  • Incorporates Charged Surface Hybrid (CSH) technology for enhanced liquid chromatography performance in acidic, low ionic strength mobile phases.
  • Offers increased loading capacity and particle size scalability, typical of fully-porous particle columns.