Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, XSelect CSH Phenyl-Hexyl, 2.5 μm, 3 mm × 150 mm, 130 Å

Catalog No. 50465936 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XSelect CSH Phenyl-Hexyl columns are designed for robust separation performance and offer alternative analyte selectivity compared to traditional reversed-phase columns. They are a valuable tool for method development and enable a seamless transition path toward UPLC adoption.

  • Alternative analyte selectivity.
  • Robust, low bleed sorbent with trifunctionally bonded C6 phenyl-ligand.
  • Selectively retains polyaromatic compounds through pi-pi interactions.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Particle size: 2.5 μm.
  • Pore size: 130 Å.
  • Inner diameter: 3 mm.
  • Length: 150 mm.
  • pH range: 1 to 11.
  • Maximum pressure: 18000 psi (1240 bar).
  • eCord enabled.
  • Units per package: 3.

Catalog No. 50-465-936 Supplier Waters Corp Supplier No. 176002896
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XSelect CSH Phenyl-Hexyl columns are designed for robust separation performance and offer alternative analyte selectivity compared to traditional reversed-phase columns. They are a valuable tool for method development and enable a seamless transition path toward UPLC adoption.

  • Alternative analyte selectivity.
  • Robust, low bleed sorbent with trifunctionally bonded C6 phenyl-ligand.
  • Selectively retains polyaromatic compounds through pi-pi interactions.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Particle size: 2.5 μm.
  • Pore size: 130 Å.
  • Inner diameter: 3 mm.
  • Length: 150 mm.
  • pH range: 1 to 11.
  • Maximum pressure: 18000 psi (1240 bar).
  • eCord enabled.
  • Units per package: 3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.