Learn More
Waters Corp Waters Column, 130Å, 2.5 μm, 4.6 mm x 150 mm
Catalog No. 50466443 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development. They feature a robust C6 phenyl-ligand for selective retention of polyaromatic compounds via π–π interactions. These 2.5 μm eXtended Performance (XP) columns ensure exceptional separation performance, robustness, and throughput for HPLC, UPLC, and UHPLC technologies. Charged Surface Hybrid (CSH) technology improves performance in acidic, low ionic strength mobile phases.
  • Alternative analyte selectivity for method development.
  • Robust C6 phenyl–ligand for selective polyaromatic compound retention.
  • Exceptional separation performance and throughput for HPLC, UPLC, and UHPLC.
  • Improved performance in acidic, low ionic strength mobile phases with CSH technology.
Catalog No. 50-466-443 Supplier Waters Corp Supplier No. 186006735
Add to Cart
Add to Cart
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development. They feature a robust C6 phenyl-ligand for selective retention of polyaromatic compounds via π–π interactions. These 2.5 μm eXtended Performance (XP) columns ensure exceptional separation performance, robustness, and throughput for HPLC, UPLC, and UHPLC technologies. Charged Surface Hybrid (CSH) technology improves performance in acidic, low ionic strength mobile phases.
  • Alternative analyte selectivity for method development.
  • Robust C6 phenyl–ligand for selective polyaromatic compound retention.
  • Exceptional separation performance and throughput for HPLC, UPLC, and UHPLC.
  • Improved performance in acidic, low ionic strength mobile phases with CSH technology.