Learn More
Waters Corp Column, Phenyl-Hexyl, 130Å, 5 μm, 4.6 mm, 50 mm
Supplier:  Waters Corp 186005397
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity compared to traditional reversed-phase columns, making them a valuable tool for method development. They feature a trifunctionally bonded C6 phenyl-ligand, which is a robust, low bleed sorbent designed to selectively retain polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity
  • Trifunctionally bonded C6 phenyl-ligand
  • Robust, low bleed sorbent
  • Selectively retains polyaromatic compounds through pi-pi interactions
Catalog No. 50-239-1708