Learn More
Waters Corp Waters Column, Phenyl-Hexyl, 3.5 μm, 4.6 mm x 100 mm, 130 Å
Catalog No. 50791398 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development. Featuring a robust, low bleed trifunctionally bonded C6 phenyl-ligand, these columns deliver exceptional separation performance and throughput across HPLC, UPLC, and UHPLC assays, with improved loading capacity and particle size scalability.
  • Alternative analyte selectivity for method development.
  • Robust, low bleed trifunctionally bonded C6 phenyl-ligand.
  • Compatible with HPLC, UPLC, and UHPLC systems.
  • Outstanding performance and equivalent backpressure compared to core-shell columns.
  • Charged Surface Hybrid (CSH) technology for acidic, low ionic strength mobile phases.
  • Non-end-capped sorbent enhances retention of acidic compounds.
  • Wide operating pH range of 1 to 11.
  • Withstands pressures up to 6,000 psi (415 Bar).
Catalog No. 50-791-398 Supplier Waters Corp Supplier No. 186005379
Waters XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development. Featuring a robust, low bleed trifunctionally bonded C6 phenyl-ligand, these columns deliver exceptional separation performance and throughput across HPLC, UPLC, and UHPLC assays, with improved loading capacity and particle size scalability.
  • Alternative analyte selectivity for method development.
  • Robust, low bleed trifunctionally bonded C6 phenyl-ligand.
  • Compatible with HPLC, UPLC, and UHPLC systems.
  • Outstanding performance and equivalent backpressure compared to core-shell columns.
  • Charged Surface Hybrid (CSH) technology for acidic, low ionic strength mobile phases.
  • Non-end-capped sorbent enhances retention of acidic compounds.
  • Wide operating pH range of 1 to 11.
  • Withstands pressures up to 6,000 psi (415 Bar).