Learn More
Waters Corp Column, CSH Phenyl-Hexyl, 5 μm, 3.0 x 30 mm
Catalog No. 50791407 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl Columns redefine HPLC performance, offering broad selectivity with enhanced reversed-phase interactions for planar aromatic compounds. CSH Technology dramatically improves LC column performance in acidic, low ionic strength mobile phases. They are designed for high analyte loading and operational stability.
  • Offers scalability across UPLC®, HPLC, and preparative platforms.
  • Provides superior peak shape for basic compounds, low column bleed, and excellent batch-to-batch reproducibility.
  • Utilizes OBD™ Packing Technology for rugged, pressure-tolerant performance and exceptional column lifetimes.
  • Supports efficient method development for isolation and purification.
  • Maintains chromatographic integrity even with formic acid mobile phases.
Catalog No. 50-791-407 Supplier Waters Corp Supplier No. 186005389
XSelect CSH Phenyl-Hexyl Columns redefine HPLC performance, offering broad selectivity with enhanced reversed-phase interactions for planar aromatic compounds. CSH Technology dramatically improves LC column performance in acidic, low ionic strength mobile phases. They are designed for high analyte loading and operational stability.
  • Offers scalability across UPLC®, HPLC, and preparative platforms.
  • Provides superior peak shape for basic compounds, low column bleed, and excellent batch-to-batch reproducibility.
  • Utilizes OBD™ Packing Technology for rugged, pressure-tolerant performance and exceptional column lifetimes.
  • Supports efficient method development for isolation and purification.
  • Maintains chromatographic integrity even with formic acid mobile phases.