Learn More
Waters Corp Waters XSelect Column, 130Å, 3.5 μm, 3 mm X 150 mm
Catalog No. 50791392 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development. They feature a robust, low-bleed C6 phenyl-ligand that selectively retains polyaromatic compounds through pi-pi interactions, focusing on reversed-phase contacts.
  • Superior performance and comparable backpressure to core-shell columns.
  • Higher loading capacity and particle size scalability.
  • Charged Surface Hybrid (CSH) technology improves performance in acidic, low ionic strength mobile phases.
  • Non-end-capped sorbent enhances acidic chemical retention.
  • Designed for high separation performance and throughput in HPLC assays.
  • Optimizes selectivity differences for method development flexibility.
  • Operates within a pH range of 1–11.
  • Tolerates a maximum pressure of 6000 psi (415 Bar).
Catalog No. 50-791-392 Supplier Waters Corp Supplier No. 186005373
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development. They feature a robust, low-bleed C6 phenyl-ligand that selectively retains polyaromatic compounds through pi-pi interactions, focusing on reversed-phase contacts.
  • Superior performance and comparable backpressure to core-shell columns.
  • Higher loading capacity and particle size scalability.
  • Charged Surface Hybrid (CSH) technology improves performance in acidic, low ionic strength mobile phases.
  • Non-end-capped sorbent enhances acidic chemical retention.
  • Designed for high separation performance and throughput in HPLC assays.
  • Optimizes selectivity differences for method development flexibility.
  • Operates within a pH range of 1–11.
  • Tolerates a maximum pressure of 6000 psi (415 Bar).