Learn More
Waters Corp Waters Corporation Column, Phenyl-Hexyl, 5 μm, 2.1 mm, 150 mm
Catalog No. 50791404 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity compared to traditional reversed-phase columns
  • Trifunctionally bonded C6 phenyl-ligand
  • Robust, low bleed sorbent
  • Selectively retains polyaromatic compounds through pi-pi interactions
  • pH range of 1 to 11
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped design
  • 5 μm spherical particle size
  • 130 Å pore size
  • 2.1 mm inner diameter and 150 mm length
Catalog No. 50-791-404 Supplier Waters Corp Supplier No. 186005386
Add to Cart
Add to Cart
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity compared to traditional reversed-phase columns
  • Trifunctionally bonded C6 phenyl-ligand
  • Robust, low bleed sorbent
  • Selectively retains polyaromatic compounds through pi-pi interactions
  • pH range of 1 to 11
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped design
  • 5 μm spherical particle size
  • 130 Å pore size
  • 2.1 mm inner diameter and 150 mm length