Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Column, 130Å, 5 μm, 2.1 mm x 100 mm
Catalog No. 50791403 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity compared to traditional reversed-phase columns, aiding method development. They feature a trifunctionally bonded C6 phenyl-ligand, a robust, low-bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity for method development
  • Trifunctionally bonded C6 phenyl-ligand, robust and low-bleed
  • Selectively retains polyaromatic compounds via pi-pi interactions
  • Emphasizes reversed-phase interactions; broad selective HPLC family
  • High analyte loading, operational stability, and full scalability
  • Excellent reproducibility and seamless transferability
  • Powerful tool for isolation and purification
  • Maintains peak shape with formic acid mobile phases
Catalog No. 50-791-403 Supplier Waters Corp Supplier No. 186005385
Add to Cart
Add to Cart
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity compared to traditional reversed-phase columns, aiding method development. They feature a trifunctionally bonded C6 phenyl-ligand, a robust, low-bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity for method development
  • Trifunctionally bonded C6 phenyl-ligand, robust and low-bleed
  • Selectively retains polyaromatic compounds via pi-pi interactions
  • Emphasizes reversed-phase interactions; broad selective HPLC family
  • High analyte loading, operational stability, and full scalability
  • Excellent reproducibility and seamless transferability
  • Powerful tool for isolation and purification
  • Maintains peak shape with formic acid mobile phases