Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Column, 130Å, 3.5 µm, 1 mm X 100 mm, 1/pk
Catalog No. 50791379 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide you with alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
Catalog No. 50-791-379 Supplier Waters Corp Supplier No. 186005360
XSelect CSH Phenyl-Hexyl columns provide you with alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.