Learn More
Waters Corp WATERS CORPORATION HPLC Column, XSelect CSH Phenyl-Hexyl, 130 Å, 3.5 μm, 2.1 mm × 150 mm
Catalog No. 50791386 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity for method development. They utilize a trifunctionally bonded C6 phenyl-ligand and Charged Surface Hybrid (CSH) technology, ensuring robust performance across HPLC, UPLC, and UHPLC applications.
  • Superior performance and comparable backpressure versus core-shell columns.
  • Higher loading capacity and particle size scalability than totally porous particle columns.
  • Focuses on reversed-phase contacts; avoids secondary Lewis-base interactions.
  • Optimizes selectivity for flexible method development, especially with low pH mobile phases.
  • Non-end-capped sorbent enhances acidic chemical retention.
  • Ultra-low dispersion hardware for superior separation, resilience, and throughput.
  • Facilitates smooth UPLC adoption.
  • Broad working pH of 1–11.
Catalog No. 50-791-386 Supplier Waters Corp Supplier No. 186005367
XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity for method development. They utilize a trifunctionally bonded C6 phenyl-ligand and Charged Surface Hybrid (CSH) technology, ensuring robust performance across HPLC, UPLC, and UHPLC applications.
  • Superior performance and comparable backpressure versus core-shell columns.
  • Higher loading capacity and particle size scalability than totally porous particle columns.
  • Focuses on reversed-phase contacts; avoids secondary Lewis-base interactions.
  • Optimizes selectivity for flexible method development, especially with low pH mobile phases.
  • Non-end-capped sorbent enhances acidic chemical retention.
  • Ultra-low dispersion hardware for superior separation, resilience, and throughput.
  • Facilitates smooth UPLC adoption.
  • Broad working pH of 1–11.