Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect CSH Phenyl-Hexyl Column, 130Å, 5 μm

Catalog No. 50791411 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity for method development. They feature a C6 phenyl ligand for selective retention of polyaromatic compounds. This 3 mm x 250 mm column, with 5.0 μm particle and 130 Å pore size, provides robust, high-throughput performance for HPLC assays.

  • Endcapping included.
  • Inner diameter: 3 mm.
  • Reversed phase mode.
  • Particle size: 5.0 μm.
  • pH range: 1 to 11.
  • Pore size: 130 Å.
  • Length: 250 mm.
  • Provides alternative analyte selectivity and robust, low-bleed performance.
  • Selectively retains polyaromatic compounds via pi-pi interactions.
  • Offers exceptional separation performance and high throughput.

Catalog No. 50-791-411 Supplier Waters Corp Supplier No. 186005393
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity for method development. They feature a C6 phenyl ligand for selective retention of polyaromatic compounds. This 3 mm x 250 mm column, with 5.0 μm particle and 130 Å pore size, provides robust, high-throughput performance for HPLC assays.

  • Endcapping included.
  • Inner diameter: 3 mm.
  • Reversed phase mode.
  • Particle size: 5.0 μm.
  • pH range: 1 to 11.
  • Pore size: 130 Å.
  • Length: 250 mm.
  • Provides alternative analyte selectivity and robust, low-bleed performance.
  • Selectively retains polyaromatic compounds via pi-pi interactions.
  • Offers exceptional separation performance and high throughput.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.