Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Column, 3.5 μm, 2.1 x 50 mm
Catalog No. 50791384 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity, valuable for method development. They feature a robust, low bleed C6 phenyl-ligand that selectively retains polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity for method development.
  • Robust, low bleed C6 phenyl-ligand for selective polyaromatic compound retention.
  • Superior performance and comparable back pressure to core-shell columns.
  • Increased loading capacity and particle size scalability.
  • CSH technology enhances LC performance in acidic, low ionic strength mobile phases.
  • Optimized for low pH selectivity; non-end-capped sorbent enhances acidic chemical retention.
  • Ultra-low dispersion hardware for remarkable separation and throughput in HPLC tests.
  • Broad operating pH range of 1–11 and maximum pressure of 6000 psi (415 Bar).
Catalog No. 50-791-384 Supplier Waters Corp Supplier No. 186005365
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity, valuable for method development. They feature a robust, low bleed C6 phenyl-ligand that selectively retains polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity for method development.
  • Robust, low bleed C6 phenyl-ligand for selective polyaromatic compound retention.
  • Superior performance and comparable back pressure to core-shell columns.
  • Increased loading capacity and particle size scalability.
  • CSH technology enhances LC performance in acidic, low ionic strength mobile phases.
  • Optimized for low pH selectivity; non-end-capped sorbent enhances acidic chemical retention.
  • Ultra-low dispersion hardware for remarkable separation and throughput in HPLC tests.
  • Broad operating pH range of 1–11 and maximum pressure of 6000 psi (415 Bar).