Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Method Validation Kit, 130Å, 3.5 μm, 3 mm x 150 mm
Catalog No. 50791519 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HPLC columns, powered by CSH Technology, offer the widest range of selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases. XSelect Method Validation Kits contain three columns, each packed with a different lot of sorbent.
  • Designed for selectivity and stability at low pH.
  • Superior peak shape for basic compounds.
  • Highest analyte mass loading available.
  • Excellent batch-to-batch reproducibility.
  • Ideal for rapid method development.
  • Recommended use: For improving selectivity, peak shape, and retentivity in methods development.
Catalog No. 50-791-519 Supplier Waters Corp Supplier No. 186005562
XSelect HPLC columns, powered by CSH Technology, offer the widest range of selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases. XSelect Method Validation Kits contain three columns, each packed with a different lot of sorbent.
  • Designed for selectivity and stability at low pH.
  • Superior peak shape for basic compounds.
  • Highest analyte mass loading available.
  • Excellent batch-to-batch reproducibility.
  • Ideal for rapid method development.
  • Recommended use: For improving selectivity, peak shape, and retentivity in methods development.