Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Method Validation Kit, 130Å, 5 µm, 2.1 mm X 150 mm, 3/pk
Supplier:  Waters Corp 186005565
Encompass_Preferred
XSelect HPLC columns, powered by CSH Technology, offer the widest range of selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases. XSelect Method Validation Kits contain three columns, each packed with a different gel lot of stationary phase material.
Catalog No. 50-791-522