Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Method Validation Kit, 130Å, 3.5 µm, 2.1 mm X 100 mm, 3/pk
Catalog No. 50791517 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HPLC columns, powered by CSH Technology, offer the widest range of selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases. XSelect Method Validation Kits contain three columns, each packed with a different gel lot of stationary phase material.
Catalog No. 50-791-517 Supplier Waters Corp Supplier No. 186005560
XSelect HPLC columns, powered by CSH Technology, offer the widest range of selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases. XSelect Method Validation Kits contain three columns, each packed with a different gel lot of stationary phase material.