Learn More
Waters Corp Waters OBD Preparative Column, CSH Phenyl-Hexyl, 5 μm, 10 x 100 mm
Catalog No. 502391092 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XSelect CSH Phenyl-Hexyl OBD Preparative Columns are designed for seamless scalability from UPLC, UHPLC, or HPLC screening to lab-scale purification. They feature CSH Technology for added selectivity and low pH loadability, ensuring consistent performance and extended column lifetimes.
  • Designed for direct scalability from UPLC™, UHPLC, or HPLC screening to lab-scale purification.
  • Utilizes Optimum Bed Density (OBD™) design for predictable, uniform density profiles, eliminating potential for voids and extending column lifetime.
  • Provides added selectivity and low pH loadability due to CSH Technology.
  • Maximizes productivity when scaling up screening methods to preparative HPLC.
Catalog No. 50-239-1092 Supplier Waters Corp Supplier No. 186008245
Add to Cart
Add to Cart
Waters XSelect CSH Phenyl-Hexyl OBD Preparative Columns are designed for seamless scalability from UPLC, UHPLC, or HPLC screening to lab-scale purification. They feature CSH Technology for added selectivity and low pH loadability, ensuring consistent performance and extended column lifetimes.
  • Designed for direct scalability from UPLC™, UHPLC, or HPLC screening to lab-scale purification.
  • Utilizes Optimum Bed Density (OBD™) design for predictable, uniform density profiles, eliminating potential for voids and extending column lifetime.
  • Provides added selectivity and low pH loadability due to CSH Technology.
  • Maximizes productivity when scaling up screening methods to preparative HPLC.