Learn More
Waters Corp VanGuard Cartridge, Phenyl-Hexyl, 5 μm, 3.9 mm × 5 mm
Catalog No. 502390951 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These VanGuard Cartridges are engineered to prolong the lifespan and enhance the performance of analytical columns. They achieve this by effectively removing particulate contamination from the mobile phase, ensuring optimal chromatographic results. This specific cartridge is tailored to safeguard analytical columns with a 4.6 mm inner diameter and 5 μm sorbent particles.
  • Removes particulates and chemical contamination.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Provides cost-effective protection for analytical columns.
  • Extends analytical column lifetime and performance.
  • Optimized to protect 4.6 mm I.D. analytical columns containing 5 μm sorbent particles.
Catalog No. 50-239-0951 Supplier Waters Corp Supplier No. 186007838
These VanGuard Cartridges are engineered to prolong the lifespan and enhance the performance of analytical columns. They achieve this by effectively removing particulate contamination from the mobile phase, ensuring optimal chromatographic results. This specific cartridge is tailored to safeguard analytical columns with a 4.6 mm inner diameter and 5 μm sorbent particles.
  • Removes particulates and chemical contamination.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Provides cost-effective protection for analytical columns.
  • Extends analytical column lifetime and performance.
  • Optimized to protect 4.6 mm I.D. analytical columns containing 5 μm sorbent particles.