Learn More
Waters Corp Waters XSelect Column, 130Å, 2.5 μm, 3 mm x 50 mm, Phenyl-Hexyl
Supplier:  Waters Corp 186006247
Encompass_Preferred
  • Phenyl chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 11
  • Maximum pressure of 18000 psi (1240 bar)
  • Endcapped
  • Phenyl-Hexyl bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • Parker-style endfitting type
  • 130 Å pore size
  • Column format
  • 185 surface area
  • Compatible with UPLC and UHPLC systems
  • CSH particle technology
  • L11 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 14% carbon load
  • 3 units per package
Catalog No. 50-179-610