Learn More
Waters Corp Waters Column, Phenyl-Hexyl, 2.5 μm, 3 mm x 75 mm, 130 Å
Catalog No. 50179275 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity compared to traditional reversed-phase columns, aiding in method development. They are packed in ultra-low dispersion hardware, offering exceptional separation performance and robustness.
  • Trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent.
  • Selectively retains polyaromatic compounds through π–π interactions.
  • 2.5 μm eXtended Performance [XP] columns.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-179-275 Supplier Waters Corp Supplier No. 176002616
These XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity compared to traditional reversed-phase columns, aiding in method development. They are packed in ultra-low dispersion hardware, offering exceptional separation performance and robustness.
  • Trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent.
  • Selectively retains polyaromatic compounds through π–π interactions.
  • 2.5 μm eXtended Performance [XP] columns.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.