Learn More
Waters Corp WATERS CORPORATION Column, Phenyl-Hexyl, 130Å, 2.5 μm, 4.6 mm X 50 mm
Catalog No. 50201547 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CSH Phenyl-Hexyl columns offer an alternative analyte selectivity for method development. They provide exceptional separation performance, robustness, and throughput for HPLC assays and facilitate a seamless transition to UPLC adoption.
  • Provides alternative analyte selectivity compared to traditional reversed-phase columns.
  • Valuable tool for method development.
  • Utilizes a robust, low bleed trifunctionally bonded C6 phenyl-ligand sorbent.
  • Selectively retains polyaromatic compounds through pi-pi interactions.
  • Packed in ultra-low dispersion hardware for eXtended Performance (XP).
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Facilitates a seamless transition path toward UPLC adoption.
Catalog No. 50-201-547 Supplier Waters Corp Supplier No. 186006133
CSH Phenyl-Hexyl columns offer an alternative analyte selectivity for method development. They provide exceptional separation performance, robustness, and throughput for HPLC assays and facilitate a seamless transition to UPLC adoption.
  • Provides alternative analyte selectivity compared to traditional reversed-phase columns.
  • Valuable tool for method development.
  • Utilizes a robust, low bleed trifunctionally bonded C6 phenyl-ligand sorbent.
  • Selectively retains polyaromatic compounds through pi-pi interactions.
  • Packed in ultra-low dispersion hardware for eXtended Performance (XP).
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Facilitates a seamless transition path toward UPLC adoption.