Learn More
Waters Corp Waters Column, CSH Phenyl-Hexyl, 2.5 μm, 2.1 mm × 50 mm
Catalog No. 50179530 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CSH Phenyl-Hexyl columns offer an alternative analyte selectivity compared to traditional reversed-phase columns, providing exceptional separation performance and robustness for HPLC assays, and enabling a seamless transition to UPLC adoption. They integrate Charged Surface Hybrid (CSH) technology for enhanced performance in acidic, low ionic strength mobile phases.
  • Provides alternative analyte selectivity.
  • Robust, low bleed sorbent with trifunctionally bonded C6 phenyl-ligand.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
  • Integrates Charged Surface Hybrid (CSH) technology for enhanced LC performance.
  • Fully compatible with all HPLC, UPLC, and UHPLC technologies.
  • Non-end-capped sorbent improves acidic compound retention.
Catalog No. 50-179-530 Supplier Waters Corp Supplier No. 186006125
Add to Cart
Add to Cart
CSH Phenyl-Hexyl columns offer an alternative analyte selectivity compared to traditional reversed-phase columns, providing exceptional separation performance and robustness for HPLC assays, and enabling a seamless transition to UPLC adoption. They integrate Charged Surface Hybrid (CSH) technology for enhanced performance in acidic, low ionic strength mobile phases.
  • Provides alternative analyte selectivity.
  • Robust, low bleed sorbent with trifunctionally bonded C6 phenyl-ligand.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
  • Integrates Charged Surface Hybrid (CSH) technology for enhanced LC performance.
  • Fully compatible with all HPLC, UPLC, and UHPLC technologies.
  • Non-end-capped sorbent improves acidic compound retention.